ABT-737 ≥95%

Supplier: 0000027161
Warning

Synonyms: ABT737, (R)-4-(4-((4'-Chloro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-(dimethylamino)-1-(phenylthio)butan-2-yl)amino)-3-nitrophenyl)sulphonyl)benzamide, (R)-4-(4-((4'-Chloro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-(dimethylamino)-1-(phenylthio)butan-2-yl)amino)-3-nitrophenyl)sulfonyl)benzamide

SYN-1001-M005 SYN-1001-M010 SYN-1001-M050 SYN-1001-M001
102991-174EA 134.31 USD
102991-174 102991-176 102991-178 102991-292
ABT-737 ≥95%
ABT-737

ABT-737 is a small-molecule inhibitor of the anti-apoptotic proteins Bcl-2, Bcl-X(L) and Bcl-w, with an affinity two to three orders of magnitude more potent than previously reported compounds. Mechanistic studies reveal that ABT-737 does not directly initiate the apoptotic process, but enhances the effects of death signals, displaying synergistic cytotoxicity with chemotherapeutics and radiation. ABT-737 exhibits single-agent-mechanism-based killing of cells from lymphoma and small-cell lung carcinoma lines, as well as primary patient-derived cells, and in animal models, ABT-737 improves survival, causes regression of established tumors, and produces cures in a high percentage of the mice. ABT-737 binds to Bcl-2, Bcl-xL and Bcl-w with very high affinities (Ki 1 nM) and also shows a very high specificity over Mcl-1 and A1.

Formula: C₄₂H₄₅ClN₆O₅S₂
Storage Temperature: Freezer
MDL Number: MFCD12756212
CAS Number: 852808-04-9

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR