1,2,3-Tris(diethylamino)cyclopropenylium bis(trifluoromethanesulfonyl)imide ≥97%

Supplier: 0000027161
Warning

CDX-T0091-G001 CDX-T0091-G005
102991-108EA 78.92 USD
102991-108 102991-110
1,2,3-Tris(diethylamino)cyclopropenylium bis(trifluoromethanesulfonyl)imide ≥97%
1,2,3-Tris(diethylamino)cyclopropenylium bis(trifluoromethanesulfonyl)imide

Air- and water-stable ionic liquid. The properties of these ionic liquids include almost zero vapour pressure, low flammability and ease of recycling. They are frequently considered as potential green alternatives to classical organic solvents. Additionally, most classes can be readily tuned to provide unique solvent environments in which to carry out reactions, extractions and other processes. There are a large number of applications.

Formula: C₁₇H₃₀F₆N₄O₄S₂
Storage Temperature: Freezer
CAS Number: 1333477-58-9

Order Now

Specification Test Results

Source/Host Synthetic
Purity 97% (HPLC)
Appearance/Color Yellow liquid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR