(±)-Sodium-2,3-dihydroxyisovalerate hydrate ≥96%

Supplier: 0000027161
Warning

Synonyms: Sodium 2,3-dihydroxy-3-methylbutanoate hydrate

CDX-S0055-M020 CDX-S0055-M100
102991-014EA 123.23 USD
102991-014 102991-016
(±)-Sodium-2,3-dihydroxyisovalerate hydrate ≥96%
(±)-Sodium-2,3-dihydroxyisovalerate hydrate

Racemic form of intermediate in L-valine catabolism and anabolism.

Formula: C₅H₉NaO₄·H₂O
Storage Temperature: Freezer
MDL Number: MFCD12912381
CAS Number: 74294-97-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR