N-(3-Succinimidyloxycarbonylphenyl)methyl-4-(5'-(4''-methoxy-phenyl)-2'-oxazolyl)pyridinium bromide ≥95%

Supplier: 0000027161
Warning

Synonyms: Stain 7-I

CDX-S0030-M005 CDX-S0030-M050 CDX-S0030-M250
102990-982EA 78.92 USD
102990-982 102990-984 102990-986
N-(3-Succinimidyloxycarbonylphenyl)methyl-4-(5'-(4''-methoxy-phenyl)-2'-oxazolyl)pyridinium bromide ≥95%
N-(3-Succinimidyloxycarbonylphenyl)methyl-4-(5'-(4''-methoxy-phenyl)-2'-oxazolyl)pyridinium bromide

New class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance.

Formula: C₂₇H₂₂BrN₃O₆
Storage Temperature: Freezer
CAS Number: 155862-95-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR