Pyridoxal hydrochloride

Supplier: 0000027161

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

CDX-P0118-G050 CDX-P0118-K001
102990-840EA 171.69 USD
102990-840 102990-842
Pyridoxal hydrochloride
Pyridoxal hydrochloride

For the labeling of amino acids and their detection in picomolar amounts.

Formula: C₈H₁₀ClNO₃
MDL Number: MFCD00012809
CAS Number: 65-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR