Propidium Iodide

Supplier: 0000027161

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide

CDX-P0023-M025 CDX-P0023-M500 CDX-P0023-G005
102990-796EA 113.54 USD
102990-796 102990-798 102990-800
Propidium Iodide
Propidium iodide

Propidium iodide is the most common red-fluorescent nuclear stain. It can also be excited using excitation filters appropriate for green-fluorescent dyes.

Formula: C₂₇H₃₄I₂N₄
MDL Number: MFCD00011921
CAS Number: 25535-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR