N-(3-Oxohexanoyl)-L-homoserine lactone ≥97%

Supplier: 0000027161

Synonyms: 3-oxo-C6-HSL

CDX-O0057-M010 CDX-O0057-M050
102990-730EA 88.62 USD
102990-730 102990-732
N-(3-Oxohexanoyl)-L-homoserine lactone ≥97%
N-(3-Oxohexanoyl)-L-homoserine lactone

N-(3-Oxohexanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. At increased populations of bacteria, localized higher concentrations of 3OC6-HSL, an endogenous ligand to transcriptional factor LuxR, leads to increased production of both the AHL synthase and proteins responsible for bioluminescence. Numerous other species of bacteria also employ 3OC6-HSL in cell-to-cell communication. An autoinducer of P. fischeri luciferase and a specfic genetic regulator that is unrelated to at least one of the enzyme systems that it induces. It acts after excretion and accumulation in the extracellular medium.

Formula: C₁₀H₁₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00171363
CAS Number: 143537-62-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR