(S)-α-Methoxy-2-naphthylacetic acid ≥97%

Supplier: 0000027161
Danger

Synonyms: (S)-2-NMA

CDX-M0213-M100 CDX-M0213-M500
102990-578EA 105.23 USD
102990-578 102990-580
(S)-α-Methoxy-2-naphthylacetic acid ≥97%
(S)-alpha-Methoxy-2-naphthylacetic acid

Excellent chiral anisotropic NMR reagent. Used for the determination of the absolute configuration of secondary alcohols.

Formula: C₁₃H₁₂O₃
Storage Temperature: Freezer
MDL Number: MFCD10000872
CAS Number: 157134-51-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic
Purity 97% (HPLC)
Appearance/Color White powder.
Solubility Soluble in methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR