Methyl-5-hydroxynicotinate ≥96%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

CDX-M0130-G001 CDX-M0130-GG25 CDX-M0130-G010
102990-524EA 138.46 USD
102990-524 102990-526 102990-528
Methyl-5-hydroxynicotinate ≥96%
Methyl 5-hydroxynicotinate

Heterocyclic building block.

Formula: C₇H₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00191521
CAS Number: 30766-22-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR