2-Methoxy-2-(2-naphthyl)ethanenitrile ≥95%

Supplier: 0000027161
Warning

Synonyms: Methoxy(2-naphthyl)acetonitrile

CDX-M0072-M050 CDX-M0072-M250
102990-430EA 202.15 USD
102990-430 102990-432
2-Methoxy-2-(2-naphthyl)ethanenitrile ≥95%
2-Methoxy-2-(2-naphthyl)ethanenitrile

Fluorescent dye.

Formula: C₁₃H₁₁NO
Storage Temperature: Freezer
CAS Number: 118736-08-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR