1-[2-(6-Methoxy-3-oxo-3H-xanthen-9-yl)benzoyl]piperidine-4-carboxylic acid ≥95%

Supplier: 0000027161
Warning

Synonyms: Xanthamide 8

CDX-M0071-M020 CDX-M0071-M125
102990-426EA 319.85 USD
102990-426 102990-428
1-[2-(6-Methoxy-3-oxo-3H-xanthen-9-yl)benzoyl]piperidine-4-carboxylic acid ≥95%
1-[2-(6-Methoxy-3-oxo-3H-xanthen-9-yl)benzoyl]piperidine-4-carboxylic acid

Fluorescent dye.

Formula: C₂₇H₂₃NO₆
Storage Temperature: Freezer
CAS Number: 442151-56-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR