N-(3-Isothiocyanatopropyl)-4-(5'-(4''-methoxyphenyl)-2'-oxazolyl) pyridinium bromide ≥90%

Supplier: 0000027161
Warning

CDX-I0036-M050 CDX-I0036-M250
102990-204EA 210.46 USD
102990-204 102990-286
N-(3-Isothiocyanatopropyl)-4-(5'-(4''-methoxyphenyl)-2'-oxazolyl) pyridinium bromide ≥90%
N-(3-Isothiocyanatopropyl)-4-(5'-(4''-methoxyphenyl)-2'-oxazolyl) pyridinium bromide

Fluorescent reagent with high selectivity.

Formula: C₁₉H₁₈BrN₃O₂S
Storage Temperature: Freezer
CAS Number: 1034443-41-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR