N-Ethanoyl-DL-homoserine lactone ≥98%
Supplier: 0000027161
|
Warning
|
Synonyms:
C2-HSL
CDX-E0071-M005
CDX-E0071-M025
102989-920EA
123.23
USD
102989-920
102989-922
N-Ethanoyl-DL-homoserine lactone ≥98%
N-Ethanoyl-DL-homoserine lactone
N-Ethanoyl-DL-homoserine lactone is the shortest alkyl homologue and most polar of a family of mediators of cell to cell interactions in bacterial biofilms. Acylhomoserine lactones have been detected in hundreds of bacterial species. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. N-Ethanoyl-DL-homoserine lactone is not found in nature and can serve as a polar negative control for quorum sensing events.
Formula:
C₆H₉NO₃ Storage Temperature: Freezer |
CAS Number:
114375-78-9 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in ethanol, methanol, DMF or DMSO. Poorly soluble in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...