Soybean oil, epoxidised ≥98%

Supplier: 0000027161
Warning

CDX-E0037-G010 CDX-E0037-G050
102989-900EA 157.85 USD
102989-900 102989-902
Soybean oil, epoxidised ≥98%
Soybean oil, epoxidised

Useful for the determination of ESBO in foods with HPLC-MS or GC-MS. ESBO is currently employed as plasticizer and HCl scavenger in PVC products. Vegetable oils are widely used for chemical manipulation because of the high numbers of carbon-carbon double bonds. Through the epoxidation, an epoxide group (which is a more reactive group than a double bond) is added to the soybean oil, transforming it in a good hydrochloric acid scavenger and good plasticizer.

Formula: C₅₇H₉₈O₁₂
Storage Temperature: Freezer
MDL Number: MFCD00163560
CAS Number: 8013-07-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (CHN)
Appearance/Color Yellow to brown oil.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR