Eosin-5-isothiocyanate ≥98%

Supplier: ADIPOGEN CORP MS
Danger

CDX-E0013-M050 CDX-E0013-M250
102989-886EA 210.46 USD
102989-886 102989-888
Eosin-5-isothiocyanate ≥98%
Eosin-5-isothiocyanate

Fluorescent probe. Long wavelength red labeling derivative for proteins. Can be used as a reversible inhibitor of the erythrocyte calcium pump as well as an effective phosphorescent probe in measurement of rotational diffusion of proteins and in fluorescence energy transfer (FRET) studies. EITC tagged molecules may be used together with fluorescein-5′-isothiocyanate (FITC) tagged molecules in fluoresce resonance energy transfer (FRET) based assays. Spectral properties: Abs/Em = 520/544 nm

Formula: C₂₁H₇Br₄NO₅S
Storage Temperature: Freezer
MDL Number: MFCD00082502
CAS Number: 60520-47-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Orange to red powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR