Dihydroxy-MEID ≥98%

Supplier: 0000027161

Synonyms: (3aR,4R,5S,6R,7S,7aS)-Hexahydro-5,6-dihydroxy-2-methyl-4,7-epoxy-1H-isoindole-1,3(2H)-dione

CDX-D0507-M050 CDX-D0507-M100
102989-844EA 157.85 USD
102989-844 102989-846
Dihydroxy-MEID ≥98%
Dihydroxy-MEID

Building block for oligonucleotide synthesis.

Formula: C₉H₁₁NO₅
Storage Temperature: Refrigerator
CAS Number: 730963-39-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White crystalline powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR