DAR-MT ≥98%

Supplier: 0000027161
Warning

Synonyms: Diaminorhodamine-M triazole

CDX-D0274-M001 CDX-D0274-M005
102989-716EA 351.69 USD
102989-716 102989-718
DAR-MT ≥98%
DAR-MT

Reference compound for the fluorescent (nitric oxide) NO probe DAR-M.

Formula: C₂₉H₃₁N₅O₃
Storage Temperature: Freezer
CAS Number: 261351-47-7

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color Dark violet powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR