N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
Supplier: 0000027161
Synonyms:
DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester
CDX-D0236-M025
CDX-D0236-M100
CDX-D0236-G001
102989-688EA
91.38
USD
102989-688
102989-690
102989-692
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
N-terminal modification of peptides in automated synthesis.
Formula:
C₁₉H₁₈N₄O₄ |
MDL Number:
MFCD00467415 CAS Number: 146998-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...