1,4-Cubanebis(dimethylamide) ≥96%
Supplier: 0000027161
Synonyms:
N1,N1,N4,N4-tetramethyl-pentacyclo[4.2.0.02,5.03,8.04,7]octane-1,4-dicarboxamide
CDX-C0238-M025
CDX-C0238-M100
102989-568EA
279.69
USD
102989-568
102989-570
1,4-Cubanebis(dimethylamide) ≥96%
1,4-Cubanebis(dimethylamide)
Building block for synthesis.
Formula:
C₁₄H₁₈N₂O₂ Storage Temperature: Refrigerator |
CAS Number:
133180-93-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...