3-Cyano-7-ethoxycoumarin

Supplier: 0000027161

CDX-C0092-M020 CDX-C0092-M100
102989-458EA 254.77 USD
102989-458 102989-460
3-Cyano-7-ethoxycoumarin
3-Cyano-7-ethoxycoumarin

Substrate for determination of cytochrom P-450 dependent oxidases; about 50-100 times more sensitive than conventional substrate ethoxy-resorufin due to higher turn over rate.

Formula: C₁₂H₉NO₃
MDL Number: MFCD00467359
CAS Number: 117620-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR