1,1-Bis(phenylsulfonyl)ethylene ≥98%
Supplier: 0000027161
Synonyms:
1,1-Bis(benzenesulphonyl)ethylene, 1,1-Bis(benzenesulfonyl)ethylene
CDX-B0127-G001
CDX-B0127-G005
102989-372EA
96.92
USD
102989-372
102989-374
1,1-Bis(phenylsulfonyl)ethylene ≥98%
1,1-Bis(phenylsulfonyl)ethylene
Synthetic compound equivalent to the ethylene 1,2-dipole. Dienophile used for Diels-Alder reactions and useful synthon for neutral homologation of ketones. Used in the preparation of alpha,alpha-disubstituted alpha-amino acid derivatives.
Formula:
C₁₄H₁₂O₄S₂ Storage Temperature: Freezer |
MDL Number:
MFCD00043149 CAS Number: 39082-53-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to faint yellow powder. |
Solubility | Soluble in toluene. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...