2-Bromo-1-ethylpyridinium tetrafluoroborate

Supplier: ADIPOGEN CORP MS

Synonyms: BEP, 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate

CDX-B0040-G001 CDX-B0040-G005 CDX-B0040-G025
102989-338EA 88.62 USD
102989-338 102989-340 102989-342
2-Bromo-1-ethylpyridinium tetrafluoroborate
2-Bromo-1-ethylpyridinium tetrafluoroborate

Efficient coupling reagent for the synthesis of sterically hindered amides, especially peptides. In terms of reactivity and racemization-suppressing capability, this pyridinium-type reagent, BEP, proved to be more efficient than commonly used uronium- and phosphonium-type reagents, such as HBTU, BOP, and PyBroP, for the synthesis of hindered peptides containing N-methylated or Calpha,Calpha-dialkylated amino acid residues. BEP was also proven to be an efficient reagent for the synthesis of esters, especially active esters and hindered esters, and tert-butyl esters.

Formula: C₇H₉BBrF₄N
MDL Number: MFCD00060046
CAS Number: 878-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR