8-Butyryloxy-pyren-1,3,6-trisulfonic acid ≥95%

Supplier: 0000027161
Warning

CDX-B0025-M050 CDX-B0025-M250
102989-318EA 257.54 USD
102989-318 102989-320
8-Butyryloxy-pyren-1,3,6-trisulfonic acid ≥95%
8-Butyryloxy-pyren-1,3,6-trisulfonic acid

Highly water soluble enzyme substrate for the fluorimetric assay of esterases at longwave excitation and emission wavelengths.

Formula: C₂₀H₁₃Na₃O₁₁S₃
Storage Temperature: Freezer
CAS Number: 115787-82-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO, DMF water or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR