(S)-(+)-1-Aminoindan

Supplier: ADIPOGEN CORP MS

Synonyms: (S)-(+)-1-Indanylamine

CDX-A0181-M250 CDX-A0181-G005 CDX-A0181-G025
102989-234EA 98.31 USD
102989-234 102989-236 102989-238
(S)-(+)-1-Aminoindan
(S)-(+)-1-Aminoindan

Amine used as chiral and organic building block.

Formula: C₉H₁₁N
MDL Number: MFCD00216670
CAS Number: 61341-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR