Actinomycin X2 (from Streptomyces species) ≥98% (by HPLC)

Supplier: 0000027161

BVT-0375-M001 BVT-0375-M005
102988-930EA 124.62 USD
102988-930 102988-932
Actinomycin X2 (from Streptomyces species) ≥98% (by HPLC)
Actinomycin X2

Actinomycin X2 is an anti-tumor and anti-viral antibiotic.


  • Apoptosis inducer
  • Forms a complex with DNA
  • Anti-tuberculosis agent.


Aniso-Actinomycin (4-Oxopro instead of Pro in the beta-chain). Has higher cytotoxicity towards cultured human leukemia (HL-60) cells than actinomycin D. Apoptosis inducer. Forms a complex with DNA and blocks its biological activity.


Orange crystals


Soluble in DMSO, methanol, acetone or chloroform

Storage Temperature: Freezer
CAS Number: 18865-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR