Hormaomycin (from Streptomyces griseoflavus) ≥98% (by HPLC)
Supplier: 0000027161
BVT-0107-C500
BVT-0107-M001
102988-888EA
346.15
USD
102988-888
102988-890
Hormaomycin (from Streptomyces griseoflavus) ≥98% (by HPLC)
Hormaomycin
Hormaomycin is a potent narrow-spectrum antibiotic against coryneform actinomycetes (MIC value 88 pM).
- Antibiotic
- Shows hormonal activity
- Rare type of natural product
Antibacterial agent (selective). Quorum sensing modulator. Inducer of morphological differentiation in actinomycetes. Stimulator of antibiotic production. Inducer of aerial mycelium formation in various actinomycetes. Antimalarial (in vitro). Rare type of natural product, regulating bacterial behavior and independently function in a defensive antibiotic role.
Formula:
C₅₅H₆₉N₁₀O₁₄Cl |
CAS Number:
121548-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...