Benadrostin ≥98%

Supplier: 0000027161
Warning

Synonyms: 8-Hydroxy-2H-1,3-benzoxazine-2,4(3H)-dione

BVT-0079-C500 BVT-0079-M001
102988-882EA 103.85 USD
102988-882 102988-884
Benadrostin ≥98%
Benadrostin

Poly(ADP-ribose) polymerase (PARP) inhibitor.

Formula: C₈H₅NO₄
Storage Temperature: Freezer
CAS Number: 117241-60-8

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces rubiguiosohelvolus Gö66.
Purity 98% (HPLC; NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, methanol or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR