DETA NONOate (NOC-18)

Supplier: 0000027161

Synonyms: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

AG-CR1-3614-M005 AG-CR1-3614-M025
102988-770EA 41.54 USD
102988-770 102988-772
DETA NONOate (NOC-18)
DETA NONOate (NOC-18)

Nitric oxide (NO) donor. The slow release of NO allows the use in experiments investigating the effects of prolonged constant NO delivery to cells and tissues. The half-life time is 20 hours at 37°C and 56 hours at 22-25°C in 0.1M phosphate buffer, pH 7.4. Liberates 2 mol of NO per mol of parent compound. Decomposition of NONOates is nearly instantaneous at pH 5.0.

Formula: C₄H₁₃N₅O₂
MDL Number: MFCD00278808
CAS Number: 146724-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR