DPVP (resveratrol analog)

Supplier: 0000027161
Warning

Synonyms: Resveratrol Analog I

AG-CR1-3604-M001
102988-738EA 166.15 USD
102988-738
DPVP (resveratrol analog)
DPVP (resveratrol analog)

Anticancer compound. Shown to inhibit progression of prostate cancer cells. Anti-obesity agent. 3T3-L1 adipocyte differentiation and FAS expression inhibitor.

Formula: C₁₅H₁₅NO₂
Storage Temperature: Freezer
CAS Number: 1462947-40-5

Order Now

Specification Test Results

Purity 98 (NMR)
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO or methanol. Insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR