D-(-)-N-Methylglucamine ≥95%

Supplier: 0000027161

Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol

AG-CN2-0413-G001 AG-CN2-0413-G005
102988-180EA 34.62 USD
102988-180 102988-182
D-(-)-N-Methylglucamine ≥95%
D(-)-N-Methylglucamine

Used as an excipient (an inert substance) in pharmaceuticals that improves solubility, drug release rate and stabilization of anionic drug substances. Counter ion used to replace sodium. It is impermeable towards cell membranes with comparable osmotic activity like sodium. Widely used in the preparation of contrast/radiopaque media in combination with iodine containing compounds, such as diatrizoate meglumine and iodipamide meglumine. Examples of meglumine-combined drugs are flunixin meglumine (used as a veterinary anti-inflammatory and analgesic) and meglumine antimoniate (antiprotozoal to treat leishmaniasis).

Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
Storage Temperature: Freezer
MDL Number: MFCD00004707
CAS Number: 6284-40-8

Order Now

Specification Test Results

Source/Host Synthetic. Amino sugar derived from sorbitol.
Purity 95% (NMR)
Appearance/Color White to off-white crystalline powder.
Solubility Soluble in DMSO or water. Slightly soluble in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR