Boromycin ≥99%
Supplier: 0000027161
|
Warning
|
AG-CN2-0166-C250
AG-CN2-0166-M001
102988-122EA
166.15
USD
102988-122
102988-124
Boromycin ≥99%
Boromycin
Boron-containing macrolide antibiotic
Source/Host: Isolated from Streptomyces sp.
Purity: >99% (TLC)
Appearance/Color: Off-white solid.
Solubility: Soluble in DMSO, ethanol or methanol.
Antibacterial. Antiprotozoal (Plasmodium, Babesia). Anti-HIV compound. Ca2+ homeostasis modulator. Potentiator of bleomycin-induced anti-tumor activity. Used as a chiral selector in capillary electrophoresis.
Storage Temperature:
Freezer |
CAS Number:
34524-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...