Geodin ≥95%

Supplier: 0000027161
Warning

Synonyms: Erdin (+)-form methyl ester

AG-CN2-0139-M001 AG-CN2-0139-M005
102988-044EA 180 USD
102988-044 102988-046
Geodin ≥95%
Geodin

Plasminogen activator inhibitor (PAI-1). Glucose uptake stimulator.

Formula: C₁₇H₁₂Cl₂O₇
Storage Temperature: Freezer
CAS Number: 427-63-4

Order Now

Specification Test Results

Source/Host Isolated from Penicillium sp. FKI-5002.
Purity 95% (HPLC)
Appearance/Color Pale yellow solid.
Solubility Soluble in DMSO, methanol, chloroform, acetone or ethyl acetate. Insoluble in water or hexane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR