11-O-Methylpseurotin A ≥95%
Supplier: 0000027161
|
Warning
|
AG-CN2-0121-C250
AG-CN2-0121-M001
102987-978EA
69.23
USD
102987-978
102987-980
11-O-Methylpseurotin A ≥95%
11-O-Methylpseurotin A
Antibacterial. Selectively inhibits a Hof1 deletion strain.
Formula:
C₂₃H₂₇NO₈ Storage Temperature: Freezer |
CAS Number:
956904-34-0 |
Specification Test Results
Source/Host | Isolated from Sporothrix sp. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO, ethanol or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...