Dihydrorhodamine 123

Supplier: 0000027161

CDX-D0134-M002 CDX-D0134-M010 CDX-D0134-M125
102987-888EA 108 USD
102987-888 102987-890 102987-892
Dihydrorhodamine 123
Dihydrorhodamine 123

Cell-permeable non-fluorescent reactive oxygen species (ROS) indicator. Passively diffuses across membranes where it is oxidized by peroxynitrite to rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence (Ex/Em wavelengths of 500 and 536 nm). Used to investigate reactive oxygen intermediates produced by human and murine phagocytes, activated rat mast cells and cultured endothelial cells. In addition, it has been used together with Fura Red calcium indicator to simultaneously measure oxidative bursts and Ca2+ fluxes in monocytes and granulocytes. Dihydrorhodamine 123 has been shown to be a more sensitive probe than H2DCFDA for detecting granulocyte respiratory bursts.

Formula: C₂₁H₁₈N₂O₃
MDL Number: MFCD04221428
CAS Number: 109244-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR