DAR-2 ≥97%

Supplier: 0000027161
Warning

Synonyms: 5,6-Diamino-rhodamine B

CDX-D0102-M001 CDX-D0102-M005 CDX-D0102-M025
102987-806EA 166.15 USD
102987-806 102987-808 102987-810
DAR-2 ≥97%
DAR-2

Sensitive NO probe, LOD of 10 nM, shows higher photostability than the classical fluorescein derivative DAF. Spectral data: lambdaex 562nm; lambdaem 579nm in 0.1 M phosphate pH 7.4.

Formula: C₂₈H₃₂N₄O₃
Storage Temperature: Freezer
CAS Number: 261351-45-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR