Citrinin ≥97%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Citrinin, yellow, (3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

AG-CN2-0101-M001 AG-CN2-0101-M005 AG-CN2-0101-M025
102987-338EA 55.38 USD
102987-338 102987-340 102987-342
Citrinin ≥97%
Citrinin

Antibiotic. Antiprotozoal. Antimicrobial. Cytotoxic and genotoxic in various mammalian cells. Mitochondrial permeability transition pore (MPTP) activator. Apoptosis inducer. ERK and JNK signaling pathways activator. Nephrotoxic mycotoxin. Suppresses NO and iNOS expression via inhibition of the JAK/STAT-1alpha and NF-kappaB signaling pathways. Tubulin polymerization and mitotic spindle assembly inhibitor. Increases reactive oxygen species (ROS). Induces cell cycle arrest at the G0/G1 and G2/M phase.

Formula: C₁₃H₁₄O₅
Storage Temperature: Freezer
MDL Number: MFCD00006912
CAS Number: 518-75-2
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Penicillium citrinum strain FKI-4836.
Purity 97% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in methanol, ethanol, methylene chloride, pyridine or dioxane. Insoluble in water, chloroform or hexane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR