White Line Inducing Principle ≥98%

Supplier: 0000027161

AG-CN2-0069-M001
102987-226EA 166.15 USD
102987-226
White Line Inducing Principle ≥98%
White line inducing principle

Lipodepsipeptide (LDP). Used as a diagnostic tool to identify tolaasin-producing bacteria pathogenic to mushrooms and responsible for brown blotch disease. Has haemolytic activity. Permeabilizes lipid vesicles and is involved in mushroom tissue alterations by forming transmembrane pores. Antimicrobial. Antibacterial. Antifungal.

Formula: C₅₄H₉₅N₉O₁₆
Storage Temperature: Freezer
CAS Number: 135096-89-8

Order Now

Specification Test Results

Source/Host Isolated from Pseudomonas sp.
Purity 98%
Appearance/Color White solid.
Solubility Soluble in DMSO, ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR