Colistin Sulphate (from Bacillus polymyxa)
Supplier: 0000027161
AG-CN2-0065-M100 AG-CN2-0065-G001 AG-CN2-0065-G005
102987-208EA 41.54 USD
102987-208 102987-210 102987-212
Colistin Sulphate (from Bacillus polymyxa)
Proteins and Peptides
Very potent polymyxin antibiotic
  • Antimicrobial
Has only one amino acid difference compared to polymyxin B. Targets the lipopolysaccharide (LPS) of the outer membrane of Gram-negative bacteria. Competitively displaces divalent cations (Ca2+ and Mg2+) from the negatively-charged phosphate groups LPS, causing disruption of the membrane. This increases the permeability of the cell envelope, leakage of cell contents, leading to apoptosis and cell death. Used against multidrug-resistant Gram-negative bacterial infections.