NTA Reagent Kit, Cytiva
Supplier: Cytiva
28-9950-43
102985-702EA 210.51 USD
102985-702
NTA Reagent Kit, Cytiva
Assays Protein Assays
Reagents for sensor chip NTA, used for His capture in His-tagged molecules in biomolecular interaction analysis.
  • Maximum convenience for His capture in Biacore systems
  • Provides ready-to-use solutions for use with sensor chip NTA
Capture of histidine-tagged molecules using it requires loading of the surface with nickel ions. After analyte injection the sample cycle is finished by regeneration of the surface, which removes the captured molecule. For these purposes, it contains ready-to-use nickel solution and regeneration solution.
Ordering information: The use of these products in Biacore systems requires an amine coupling Kit with sensor chip CM5 CM4 CM3 or C1. Not for use with Biacore flexchip.
Delivery information: It includes 0.5 mm NiCl₂, 50 ml, and 350 mm EDTA, 100 ml.