Anti-CD273 Monoclonal Antibody [clone: ANC8D12]

Supplier: Adipogen
ANC-273-020
102981-792EA 350.31 USD
102981-792
Anti-CD273 Monoclonal Antibody [clone: ANC8D12]
Antibodies
CD273 (B7-DC; PD-L2) is a type I surface molecule with homology to CD80, CD86, CD274. It is expressed primarily by dendritic cells and provides a stimulatory signal to CD279 (PD-1; Programmed Cell Death 1) which has an important immunoregulatory role by downregulating the T cell response. CD273 binds to CD279 (PD-1) with a 2-6 fold higher affinity than CD274 and is an important component of immune regulation.

Type: Primary
Antigen: CD273
Clonality: Monoclonal
Clone: ANC8D12
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR