Anti-CD271 Monoclonal Antibody (FITC) [clone: ANC271/3D7]

Supplier: Adipogen
ANC-271-040
102981-788EA 483.23 USD
102981-788
Anti-CD271 Monoclonal Antibody (FITC) [clone: ANC271/3D7]
Antibodies
The p75 low affinity human Nerve Growth Factor receptor CD271(NGRF, TNFRSF16) is expressed on a variety of tissue types including a subset of splenic and nodal lymphocytes. Presence of this receptor supports uptake of intracellular calcium, but not mobilization. CD271 is a stem cell marker for hematopoietic, stromal as well as cancer cells. CD271 has been reported to inhibit differentiation of mesenchymal cells into various lineages.

Type: Primary
Antigen: CD271
Clonality: Monoclonal
Clone: ANC271/3D7
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR