Anti-CD66b Monoclonal Antibody [clone: ANC3C5]

Supplier: Adipogen
ANC-219-020
102981-764EA 350.31 USD
102981-764
Anti-CD66b Monoclonal Antibody [clone: ANC3C5]
Antibodies
Human CD66b (CEACAM8, CGMb, NCA-95, CD67) is a 95kD member of the Carcinoembryonic Antigen (CEA) group. It is expressed on granulocytes and functions as an adhesion molecule. Similar to the CD66 a, c, and d isoforms, engagement of CD66b on neutrophils causes increased adhesion to endothelial cells. Crosslinking CD66b on neutrophils with antibodies triggers IL-8 release.

Type: Primary
Antigen: CD66b
Clonality: Monoclonal
Clone: ANC3C5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR