Anti-CD11C Monoclonal Antibody (APC) [clone: 3.9]

Supplier: Adipogen
ANC-160-060
102981-756EA 530.31 USD
102981-756
Anti-CD11C Monoclonal Antibody (APC) [clone: 3.9]
Antibodies
Human CD11c (alpha X integrin) complexes with CD18 (beta 2 integrin) to form the complement receptor type 4 (CR4) heterodimer which binds to fibrinogen and is involved with monocyte/granulocyte adhesion during inflammatory responses. CD11c expression is restricted to leukocytes mainly of myeloid lineage with highest expression on macrophages.

Type: Primary
Antigen: CD11C
Clonality: Monoclonal
Clone: 3.9
Conjugation: APC
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR