Anti-MG Monoclonal Antibody [clone: 3C]

Supplier: Adipogen
JAI-MMG-030N
102981-716PK 1192.15 USD
102981-716
Anti-MG Monoclonal Antibody [clone: 3C]
Antibodies
Methylglyoxal (MG), an endogenous metabolite that increases in diabetes and is a common intermediate in the Maillard reaction (glycation), reacts with proteins and forms advanced glycation end products (AGE). MG reacts with arginine residue in protein and forms numerous numbers of adducts, such as argpyrimidine.

Type: Primary
Antigen: MG
Clonality: Monoclonal
Clone: 3C
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: All, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR