Anti-4-HNE Mouse Monoclonal Antibody [clone: HNEJ-2]

Supplier: Adipogen
JAI-MHN-020P
102981-710EA 258.92 USD
102981-710
Anti-4-HNE Mouse Monoclonal Antibody [clone: HNEJ-2]
Antibodies
Reactive oxygen species (ROS) are involved in lipid peroxidation inside the living bodies, and membrane lipids are one of the major targets of ROS. During the peroxidation process, a variery of aldehydes are formed. 4-hydroxy-2-nonenal (4-HNE) is an alpha, beta unsaturated aldehyde that can be formed by peroxidation of omega-6 unsaturated fatty acids such as linoleic acid and arachidonic acid. Especially, 4-HNE formed inside living body is reported to originate from phospholipid-bound arachidonic acid. 4-HNE may be one of the most reliable biomarker of lipid peroxidation.

Type: Primary
Antigen: 4-HNE
Clonality: Monoclonal
Clone: HNEJ-2
Conjugation: Unconjugated
Epitope:
Host: Ascites
Isotype: Mouse IgG1kappa
Reactivity: All, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR