Z-Leu-Leu-Leu-B(OH)2 [MG-262]
Supplier: 0000027161
AG-CP3-0024-C200 AG-CP3-0024-M001
102981-172EA 96.92 USD
102981-172 102981-174
Z-Leu-Leu-Leu-B(OH)2 [MG-262]
Proteins and Peptides
Highly potent and selective cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Similar structure as MG-132 (AG-CP3-0011) but much higher potency.
  • Soluble in DMSO.