Human Recombinant PLGF (from Insect Cells (Sf9))
Supplier: 0000027161
AG-40T-0047-C005 AG-40T-0047-C020
102980-408EA 456.92 USD
102980-408 102980-410
Human Recombinant PLGF (from Insect Cells (Sf9))
Proteins and Peptides
Human placenta growth factor-1 (PLGF-1; PLGF) binds to the receptor FLT1/VEGFR-1. It acts as a potent chemoattractant for monocytes and only as a very weak mitogen for some endothelial cell types. The physiological function in vivo is still controversial. In several reports it was shown not to be a potent mitogen for endothelial cells and not angiogenic in vivo by using different assays. It was shown that PLGF from cell culture supernatants was angiogenic in the CAM assay and in the rabbit cornea assay.