Mouse Recombinant CREB-binding Protein (active) (from Insect cells (Sf21))
Supplier: 0000027161
AG-40T-0016-C002
102980-368EA 304.62 USD
102980-368
Mouse Recombinant CREB-binding Protein (active) (from Insect cells (Sf21))
Proteins and Peptides
CREB-binding protein acetylates histones, giving a specific tag for transcriptional activation. It also acetylates non-histone proteins, like NCOA3 and FOXO1. It binds specifically to phosphorylated CREB and enhances its transcriptional activity toward cAMP-responsive genes. Acts as a coactivator of ALX1 in the presence of EP300.