Anti-IL-1R beta Recombinant Antibody [clone: Praxy-1-1]

Supplier: Adipogen
AG-27B-0011-C100
102980-002EA 415.38 USD
102980-002
Anti-IL-1R beta Recombinant Antibody [clone: Praxy-1-1]
Antibodies
The inflammatory actions of IL-1alpha and beta depend on their interaction with its receptor, IL-1R1, and can be negatively controlled by several inhibitors such as IL-1Ra, IL-1R type 2 (IL-1R2), SIGIRR and the soluble form of IL-1 receptor accessory protein (IL-1RAcP). IL-1R2 neutralizes IL-1 either as a surface decoy receptor or in a cleaved and secreted form. IL-1R2 is expressed on B cells, on neutrophils and on in vitro expanded human Tregs.

Type: Primary
Antigen: IL-1R beta
Clonality: Recombinant
Clone: Praxy-1-1
Conjugation: Unconjugated
Epitope:
Host: Purified from HEK 293 cell culture supernatant.
Isotype: Mouse IgG2blambda
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR