Human CD273 (from CHO cells)
Supplier: 0000027161
ANC-573-020
102979-342EA 617.54 USD
102979-342
Human CD273 (from CHO cells)
Proteins and Peptides
CD273 (B7-DC; PD-L2) is a type I surface molecule with homology to CD80, CD86, CD274. It is expressed primarily by dendritic cells and provides a stimulatory signal to CD279 (PD-1; programmed death molecule) which has an important immunoregulatory role by downregulating the T cell response. CD273 binds to CD279 (PD-1) with a 2-6 fold higher affinity than CD274.